Anti-CCDC139 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-7102R-CY3
BOSSBS-7102R-CY3EA 900 EUR
BOSSBS-7102R-CY3
Anti-CCDC139 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Pseudouridination is the isomerization of uridine to pseudouridine. It is the most common posttranscriptional nucleotide modification found in RNA. It is essential for biologic functions such as spliceosome biogenesis. Pseudouridylate synthases, such as PUS10, catalyze pseudouridination of structural RNAs, including transfer, ribosomal, and splicing RNAs. These enzymes also act as RNA chaperones which facilitate the correct folding and assembly of tRNAs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC139
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR