Anti-DOCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-7110R-A555
BOSSBS-7110R-A555EA 893 EUR
BOSSBS-7110R-A555
Anti-DOCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. Functions as a guanine nucleotide exchange factor (GEF), which activates Rac Rho small GTPases by exchanging bound GDP for free GTP. Its GEF activity may be enhanced by ELMO1.Tissue specificity: Highly expressed in placenta, lung, kidney, pancreas and ovary. Expressed at intermediate level in thymus, testes and colon.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DOCK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR