Anti-Defensin alpha 6 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7129R
BOSSBS-7129REA 1090 EUR
BOSSBS-7129R
Anti-Defensin alpha 6 Rabbit Polyclonal Antibody
Antikörper
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Defensin alpha 6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR