Anti-Defensin alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7129R-A680
BOSSBS-7129R-A680EA 1350 EUR
BOSSBS-7129R-A680
Anti-Defensin alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.
Type: Primary
Antigen: Defensin alpha 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 31-100/100
Host: Rabbit
Isotype: IgG
Reactivity: Rat