Anti-Defensin alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-7129R-A750
BOSSBS-7129R-A750EA 1350 EUR
BOSSBS-7129R-A750
Anti-Defensin alpha 6 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.

Type: Primary
Antigen: Defensin alpha 6
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-100/100
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR