Anti-DEFA6 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-7129R-CY5
BOSSBS-7129R-CY5EA 1350 EUR
BOSSBS-7129R-CY5
Anti-DEFA6 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DEFA6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat