Anti-DEFA6 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-7129R-CY5.5
BOSSBS-7129R-CY5.5EA 1350 EUR
BOSSBS-7129R-CY5.5
Anti-DEFA6 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Has very low antimicrobial activity against Gram-negative and Gram-positive bacteria. May protect cells against infection with HIV-1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DEFA6
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR