Anti-CATSPER3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7136R-A680
BOSSBS-7136R-A680EA 893 EUR
BOSSBS-7136R-A680
Anti-CATSPER3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
CatSpers (cation channel, sperm associated proteins) are ion transport proteins located on the surface of sperm cells in the principal piece of the sperm tail. CatSpers are vital to sperm motility, fertilisation and cAMP-mediated calcium influx in sperm. There are four CatSper proteins in mammalian sperm, namely CatSper (or CatSper1), CatSper2, CatSper3 and CatSper4. CatSper proteins contain a single, six-transmembrane-spanning segment and exhibit the voltage-dependent Ca2+ channel four-repeat structure. CatSper proteins are believed to assemble into a heterotetrameric complex, forming anMore Product Information