Anti-ARHGEF18 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-7144R-CY3
BOSSBS-7144R-CY3EA 900 EUR
BOSSBS-7144R-CY3
Anti-ARHGEF18 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Oct 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARHGEF18
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat