Anti-MUC5AC Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-7166R-CY7
BOSSBS-7166R-CY7EA 922 EUR
BOSSBS-7166R-CY7
Anti-MUC5AC Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Gel-forming glycoprotein of gastric and respiratoy tract epithelia that protects the mucosa from infection and chemical damage by binding to inhaled microrganisms and particles that are subsequently removed by the mucocilary system.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MUC5AC
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR