Anti-SP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-7342R-A750
BOSSBS-7342R-A750EA 893 EUR
BOSSBS-7342R-A750
Anti-SP3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The Sp transcription factor family includes Sp1, Sp2, Sp3 (SPR-2) and Sp4 (SPR-1). Sp transcription factors share similar structures but do not share simi-lar functions. All four proteins contain a highly conserved DNA-binding domain composed of three zinc fingers at the C-terminus. Sp family members bind the consensus sequence GGGGCGGGGC and other closely related sequences which are known as GC boxes. Sp1, Sp3 and Sp4 share a high affinity for GC boxes while Sp2 does not. Sp2 only weakly binds to GT boxes. Sp1, Sp2 and Sp3 are ubiquitously expressed, while Sp4 is abundantly expressed inMore Product Information