Anti-ADGRB1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-7513R-CY3
BOSSBS-7513R-CY3EA 900 EUR
BOSSBS-7513R-CY3
Anti-ADGRB1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
BAI 1 protein is likely to be a potent inhibitor of angiogenesis in brain and may play a significant role as a mediator of the p53 signal in suppression of glioblastoma. It may function in cell adhesion and signal transduction in the brain. Reduced or no expression is observed in some glioblastoma cell lines and cancer tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADGRB1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR