Anti-SERCA1 ATPase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7539R-A680
BOSSBS-7539R-A680EA 893 EUR
BOSSBS-7539R-A680
Anti-SERCA1 ATPase Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
This magnesium-dependent enzyme catalyzes the hydrolysis of ATP coupled with the translocation of calcium from the cytosol to the sarcoplasmic reticulum lumen. Contributes to calcium sequestration involved in muscular excitation/contraction.
Type: Primary
Antigen: SERCA1 ATPase
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/1001
Host: Rabbit
Isotype: IgG
Reactivity: Rat