Anti-SCP1 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7542R
BOSSBS-7542REA 719 EUR
BOSSBS-7542R
Anti-SCP1 Rabbit Polyclonal Antibody
Antikörper
May be involved in vascular wall and kidney homeostasis.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR