Anti-TNIP3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-7551R-A555
BOSSBS-7551R-A555EA 893 EUR
BOSSBS-7551R-A555
Anti-TNIP3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
ABIN-3 is a member of the A20-binding inhibitor of NF-kappaB activation (ABIN) protein family. Similar to the previously characterized human ABINs (ABIN-1 and ABIN-2), ABIN-3 can bind to A20 and inhibit NF-kappaB activation. In contrast, mouse ABIN-3 is incapable of inhibiting NF-kappaB activation by proinflammatory stimuli because the protein lacks a complete ABIN homology domain, which is required for the funcitonal activity of human ABIN-3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TNIP3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR