Anti-TNFAIP3 interacting protein 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7551R-A680
BOSSBS-7551R-A680EA 893 EUR
BOSSBS-7551R-A680
Anti-TNFAIP3 interacting protein 3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
ABIN-3 is a member of the A20-binding inhibitor of NF-kappaB activation (ABIN) protein family. Similar to the previously characterised human ABINs (ABIN-1 and ABIN-2), ABIN-3 can bind to A20 and inhibit NF-kappaB activation. In contrast, mouse ABIN-3 is incapable of inhibiting NF-kappaB activation by proinflammatory stimuli because the protein lacks a complete ABIN homology domain, which is required for the funcitonal activity of human ABIN-3.
Type: Primary
Antigen: TNFAIP3 interacting protein 3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/325
Host: Rabbit
Isotype: IgG
Reactivity: Mouse