Anti-EPHA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-7554R-A555
BOSSBS-7554R-A555EA 922 EUR
BOSSBS-7554R-A555
Anti-EPHA2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Eph receptor A2 is the receptor for members of the ephrin-A family. It binds to ephrin-A1, -A3, -A4 and -A5. Eph receptor A3 is also a receptor for members of the ephrin-A family. It binds to ephrin-A2, -A3, -A4 and -A5 and is thought to play a role in lymphoid function. Eph receptor A4 is a receptor for members of the ephrin-A family. It binds to ephrin-A1, -A4 and -A5. It binds more poorly to ephrin-A2 and -A3. It may play a role in a signal transduction process involved in hindbrain pattern formation.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EPHA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat