Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-7557R-A750
BOSSBS-7557R-A750EA 1350 EUR
BOSSBS-7557R-A750
Anti-IKK alpha/beta Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta). IKK alpha and IKK beta interact with each other and both are essential for NFkB activation.
Type: Primary
Antigen: IKK alpha / beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/756
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat