Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-7557R-FITC
BOSSBS-7557R-FITCEA 1350 EUR
BOSSBS-7557R-FITC
Anti-IKK alpha + IKK beta Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
IKK is a serine protein kinase, and the IKK complex contains alpha and beta subunits (IKK alpha and IKK beta). IKK alpha and IKK beta interact with each other and both are essential for NFkB activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IKK alpha + IKK beta
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR