Anti-AZU1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-7564R-A555
BOSSBS-7564R-A555EA 893 EUR
BOSSBS-7564R-A555
Anti-AZU1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
This is a neutrophil granule-derived antibacterial and monocyte- and fibroblast-specific chemotactic glycoprotein. Binds heparin. The cytotoxic action is limited to many species of Gram-negative bacteria; this specificity may be explained by a strong affinity of the very basic N-terminal half for the negatively charged lipopolysaccharides that are unique to the Gram-negative bacterial outer envelope. It may play a role in mediating recruitment of monocytes in the second wave of inflammation. Has antibacterial activity against the Gram-nagative bacterium P.aeruginosa, this activity is inhibited by LPS from P.aeruginosa.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AZU1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human