Anti-AZU1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-7564R-CY7
BOSSBS-7564R-CY7EA 900 EUR
BOSSBS-7564R-CY7
Anti-AZU1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
This is a neutrophil granule-derived antibacterial and monocyte- and fibroblast-specific chemotactic glycoprotein. Binds heparin. The cytotoxic action is limited to many species of Gram-negative bacteria; this specificity may be explained by a strong affinity of the very basic N-terminal half for the negatively charged lipopolysaccharides that are unique to the Gram-negative bacterial outer envelope. It may play a role in mediating recruitment of monocytes in the second wave of inflammation. Has antibacterial activity against the Gram-nagative bacterium P.aeruginosa, this activity is inhibited by LPS from P.aeruginosa.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AZU1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR