Anti-A4GALT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-7588R-A488
BOSSBS-7588R-A488EA 893 EUR
BOSSBS-7588R-A488
Anti-A4GALT Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat