Anti-A4GALT Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7588R-CY5
BOSSBS-7588R-CY5EA 900 EUR
BOSSBS-7588R-CY5
Anti-A4GALT Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR