Anti-A4GALT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Lieferant: Bioss
BS-7588R-FITC
BOSSBS-7588R-FITCEA 900 EUR
BOSSBS-7588R-FITC
Anti-A4GALT Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat