Anti-A4GALT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BS-7588R-HRP
BOSSBS-7588R-HRPEA 900 EUR
BOSSBS-7588R-HRP
Anti-A4GALT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Necessary for the biosynthesis of the Pk antigen of blood histogroup P. Catalyzes the transfer of galactose to lactosylceramide and galactosylceramide. Necessary for the synthesis of the receptor for bacterial verotoxins.
Recommended Dilutions: IHC-P: 1:100-500
Type: Primary
Antigen: A4GALT
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat