Anti-GLA Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7593R
BOSSBS-7593REA 725 EUR
BOSSBS-7593R
Anti-GLA Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Galactosidase alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR