Anti-ETHE1 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-7610R-CY3
BOSSBS-7610R-CY3EA 900 EUR
BOSSBS-7610R-CY3
Anti-ETHE1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Probably plays an important role in metabolic homeostasis in mitochondria. May function as a nuclear-cytoplasmic shuttling protein that binds transcription factor RELA/NFKB3 in the nucleus and exports it to the cytoplasm. Suppresses p53-induced apoptosis by preventing nuclear localization of RELA.Involvement in disease:Defects in ETHE1 are a cause of ethylmalonic encephalopathy (EE) . EE is an autosomal recessive disorder characterized by neurodevelopmental delay and regression, recurrent petechiae, acrocyanosis, diarrhea, leading to death in the first decade of life. It is also associated with persistent lactic acidemia and ethylmalonic and methylsuccinic aciduria.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ETHE1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR