Anti-NOC2L Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-7618R-A647
BOSSBS-7618R-A647EA 893 EUR
BOSSBS-7618R-A647
Anti-NOC2L Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
NOC2L is a transcriptional corepressor with inhibitor of histone acetyltransferase (INHAT) activity. NOC2L or NIR (Novel INHAT Repressor) is ubiquitously expressed throughout embryonic development and adulthood. It is a potent transcriptional corepressor that is not blocked by histone deacetylase inhibitors and is capable of silencing both basal and activator-driven transcription. NOC2L directly binds to nucleosomes and core histones and prevents acetylation by histone acetyltransferases, thus acting as a bona fide INHAT.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NOC2L
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat