Anti-ZDHHC16 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7677R-CY5
BOSSBS-7677R-CY5EA 900 EUR
BOSSBS-7677R-CY5
Anti-ZDHHC16 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZDHHC16
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR