Anti-BCCIP Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7679R-CY5
BOSSBS-7679R-CY5EA 893 EUR
BOSSBS-7679R-CY5
Anti-BCCIP Rabbit Polyclonal Antibody (Cy5®)
Antikörper
FunctionMay promote cell cycle arrest by enhancing the inhibition of CDK2 activity by CDKN1A. May be required for repair of DNA damage by homologous recombination in conjunction with BRCA2. May not be involved in non-homologous end joining.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCCIP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR