Anti-BCCIP Rabbit Polyclonal Antibody (Cy7®)
Lieferant: Bioss
BS-7679R-CY7
BOSSBS-7679R-CY7EA 893 EUR
BOSSBS-7679R-CY7
Anti-BCCIP Rabbit Polyclonal Antibody (Cy7®)
Antikörper
FunctionMay promote cell cycle arrest by enhancing the inhibition of CDK2 activity by CDKN1A. May be required for repair of DNA damage by homologous recombination in conjunction with BRCA2. May not be involved in non-homologous end joining.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BCCIP
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat