Anti-Renalase Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7693R
BOSSBS-7693REA 719 EUR
BOSSBS-7693R
Anti-Renalase Rabbit Polyclonal Antibody
Antikörper
Catalyzes the oxidation of the less abundant alpha-NAD(P)H isoform to form beta-NAD(P)(+). The enzyme hormone is secreted by the kidney, and circulates in blood and modulates cardiac function and systemic blood pressure. Lowers blood pressure in vivo by decreasing cardiac contractility and heart rate and preventing a compensatory increase in peripheral vascular tone, suggesting a causal link to the increased plasma catecholamine and heightened cardiovascular risk. High concentrations of catecholamines activate plasma renalase and promotes its secretion and synthesis.

WB(1:100-1000)

Type: Primary
Antigen: Renalase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR