Anti-SA2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-7725R-A488
BOSSBS-7725R-A488EA 893 EUR
BOSSBS-7725R-A488
Anti-SA2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Component of cohesin complex, a complex required for the cohesion of sister chromatids after DNA replication. The cohesin complex apparently forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SA2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat