Anti-NCAPD3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-7734R-HRP
BOSSBS-7734R-HRPEA 900 EUR
BOSSBS-7734R-HRP
Anti-NCAPD3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 and SMC4. hCAP-D3 is a regulatory non-SMC subunit of the condensin II complex.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NCAPD3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR