Anti-CFAP53 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-7740R-A647
BOSSBS-7740R-A647EA 893 EUR
BOSSBS-7740R-A647
Anti-CFAP53 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
CCDC11 is a 514 amino acid protein encode by a gene that maps to human chromosome 18q21.1. Encoding over 300 genes, chromosome 18 contains about 76 million bases. Trisomy 18, or Edwards syndrome, is the second most common trisomy after Downs syndrome. Symptoms of Edwards syndrome include low birth weight, a variety of physical development defects, heart deformations and breathing difficulty. Translocation between chromosome 18 and 14 is the most common translocation in cancers, and occurs in follicular lymphomas. Niemann-Pick disease, hereditary hemorrhagic telangiectasia and erythropoietic protoporphyria are associated with chromosome 18. The TGF∫ modulators, Smad2, Smad4 and Smad7 are encoded by chromosome 18.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CFAP53
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR