Anti-HAUS7 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7776R
BOSSBS-7776REA 725 EUR
BOSSBS-7776R
Anti-HAUS7 Rabbit Polyclonal Antibody
Antikörper
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.Tissue specificity:Detected in spleen, thymus, testis, ovary, small intestine and colon, with highest levels of expression in testis and ovary.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: UCHL5IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR