Anti-CIRBP Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7790R-CY5
BOSSBS-7790R-CY5EA 893 EUR
BOSSBS-7790R-CY5
Anti-CIRBP Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Cold-inducible mRNA binding protein that plays a protective role in the genotoxic stress response by stabilizing transcripts of genes involved in cell survival. Acts as a translational activator. Seems to play an essential role in cold-induced suppression of cell proliferation. Binds specifically to the 3'-untranslated regions (3'-UTRs) of stress-responsive transcripts RPA2 and TXN. Acts as a translational repressor (By similarity). Promotes assembly of stress granules (SGs), when overexpressed.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CIRBP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR