Anti-GLP2R Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7800R
BOSSBS-7800REA 725 EUR
BOSSBS-7800R
Anti-GLP2R Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GLP2R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR