Anti-INSC Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-7838R-A350
BOSSBS-7838R-A350EA 893 EUR
BOSSBS-7838R-A350
Anti-INSC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
May function as an adapter linking the Par3 complex to the GPSM1/GPSM2 complex. Involved in spindle orientation during mitosis it may regulate cell proliferation and differentiation in the developing nervous system. May play a role in the asymmetric division of fibroblasts and participate in the process of stratification of the squamous epithelium.Tissue specificity:Isoform 1 is expressed in various tissues with stronger expression in liver, kidney and small intestine. Isoform 2 is abundantly expressed in small intestine and to a lower extent in lung and pancreas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: INSC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR