Anti-INSC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-7838R-A680
BOSSBS-7838R-A680EA 893 EUR
BOSSBS-7838R-A680
Anti-INSC Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
May function as an adapter linking the Par3 complex to the GPSM1/GPSM2 complex. Involved in spindle orientation during mitosis it may regulate cell proliferation and differentiation in the developing nervous system. May play a role in the asymmetric division of fibroblasts and participate in the process of stratification of the squamous epithelium.Tissue specificity:Isoform 1 is expressed in various tissues with stronger expression in liver, kidney and small intestine. Isoform 2 is abundantly expressed in small intestine and to a lower extent in lung and pancreas.
Type: Primary
Antigen: INSC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-300/579
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen