Anti-GAK protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-7912R-A750
BOSSBS-7912R-A750EA 893 EUR
BOSSBS-7912R-A750
Anti-GAK protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Associates with cyclin G and CDK5. Seems to act as an auxilin homolog that is involved in the uncoating of clathrin-coated vesicles by Hsc70 in non-neuronal cells. Expression oscillates slightly during the cell cycle, peaking at G1.Tissue specificity: Ubiquitous. Highest in testis.

Type: Primary
Antigen: GAK protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 244-330/1311
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR