Anti-PLK5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-7918R-A350
BOSSBS-7918R-A350EA 893 EUR
BOSSBS-7918R-A350
Anti-PLK5 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
PLK5 (Serine/threonine-protein kinase PLK5) belongs to the protein kinase superfamily and Ser/Thr protein kinase family and CDC5/Polo subfamily. PLK5P contains 1 POLO box domain and 1 Serine/Threonine protein kinase catalytic domain.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PLK5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat