Anti-PLK5 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-7918R-CY5.5
BOSSBS-7918R-CY5.5EA 900 EUR
BOSSBS-7918R-CY5.5
Anti-PLK5 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
PLK5 (Serine/threonine-protein kinase PLK5) belongs to the protein kinase superfamily and Ser/Thr protein kinase family and CDC5/Polo subfamily. PLK5P contains 1 POLO box domain and 1 Serine/Threonine protein kinase catalytic domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PLK5
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR