Anti-SHOC2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss
BS-7934R-CY3
BOSSBS-7934R-CY3EA 893 EUR
BOSSBS-7934R-CY3
Anti-SHOC2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Regulatory subunit of protein phosphatase 1 (PP1c) that acts as a M-Ras/MRAS effector and participates in MAPK pathway activation. Upon M-Ras/MRAS activation, targets PP1c to specifically dephosphorylate the 'Ser-259' inhibitory site of RAF1 kinase and stimulate RAF1 activity at specialized signaling complexes.Involvement in disease:Defects in SHOC2 are the cause of Noonan syndrome-like with loose anagen hair (NSLAH) . NSLAH children display macrocephaly, high forehead, hypertelorism, palpebral ptosis, low-set and posteriorly rotated ears, short and webbed neck and pectus anomalies. Affected subjects also have easily pluckable, sparse, thin and slow-growing hair.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SHOC2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR