Anti-ABCF3 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-7941R
BOSSBS-7941REA 725 EUR
BOSSBS-7941R
Anti-ABCF3 Rabbit Polyclonal Antibody
Antikörper
ABCF3 belongs to the ABC transporter superfamily. ABCF family. EF3 subfamily. Contains 2 ABC transporter domains. There are 2 named isoforms produced by alternative splicing.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: ABCF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR