Anti-ABHD4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Lieferant: Bioss
BS-7948R-A647
BOSSBS-7948R-A647EA 893 EUR
BOSSBS-7948R-A647
Anti-ABHD4 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Lysophospholipase selective for N-acyl phosphatidylethanolamine (NAPE). Contributes to the biosynthesis of N-acyl ethanolamines, including the endocannabinoid anandamide by hydrolyzing the sn-1 and sn-2 acyl chains from N-acyl phosphatidylethanolamine (NAPE) generating glyceroN-acyl ethanolamine (GP-NAE), an intermediate for N-acyl ethanolamine biosynthesis. Hydrolyzes substrates bearing saturated, monounsaturated, polyunsaturated N-acyl chains. Shows no significant activity towards other lysophospholipids, including lysophosphatidylcholine, lysophosphatidylethanolamine and lysophosphatidylserine.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABHD4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat