Anti-ACBD4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BS-7950R-A488
BOSSBS-7950R-A488EA 893 EUR
BOSSBS-7950R-A488
Anti-ACBD4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. ACBD4 contains 1 ACB (acyl-CoA-binding) domain. There are three isoforms. Its function is unknown.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ACBD4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat