Anti-ACBD4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-7950R-A750
BOSSBS-7950R-A750EA 893 EUR
BOSSBS-7950R-A750
Anti-ACBD4 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. ACBD4 contains 1 ACB (acyl-CoA-binding) domain. There are three isoforms. Its function is unknown.

Type: Primary
Antigen: ACBD4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-110/268
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR