Anti-ACBD4 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-7950R-CY5
BOSSBS-7950R-CY5EA 900 EUR
BOSSBS-7950R-CY5
Anti-ACBD4 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
ACBD4 is a member of the acyl-coenzyme A binding domain containing protein family. ACBD4 contains 1 ACB (acyl-CoA-binding) domain. There are three isoforms. Its function is unknown.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACBD4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR