Anti-ARMC3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-7969R-CY7
BOSSBS-7969R-CY7EA 900 EUR
BOSSBS-7969R-CY7
Anti-ARMC3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
ARMC3 contains 12 ARM repeats. ARM (Armadillo/beta-catenin-like) domains are imperfect 45-amino acid repeats involved in protein-protein interactions. ARM domain-containing proteins, such as ARMC3, function in signal transduction, development, cell adhesion and mobility, and tumor initiation and metastasis. The specific function of ARMC3 is not yet known.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARMC3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

TECHNISCHE DATEN

Antigen-Symbol ARMC3
Environmentally Preferable
Antigen-Name Armadillo Repeat Containing 3
Antigen-Synonyme KU-CT-1|CT81|Beta-catenin-like protein|ARMC3|Armadillo repeat-containing protein 3|Cancer/testis antigen 81
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Cy7®
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 219681
Isotyp IgG
ImmunoChemie Yes
Durchflusszytometrie Yes
Immunogen 760-805/872
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at –20 °C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR